4-((3-Bromobenzyl)oxy)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

544657-5G 544657-1G 544657-25G 544657-500MG
77630-260EA 1484.23 USD
77630-260 77639-586 77656-474 77651-582
4-((3-Bromobenzyl)oxy)benzonitrile ≥97%
4-((3-Bromobenzyl)oxy)benzonitrile

Precaución: For Research Use Only

Formula: C₁₄H₁₀BrNO
Storage Temperature: Ambient
CAS Number: 831229-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR