1-Bromo-2-methoxy-3,4-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

54429-5G 54429-25G 54429-1G 54429-500MG
77630-258EA 1841.64 USD
77630-258 77636-092 77639-582 77647-582
1-Bromo-2-methoxy-3,4-dimethylbenzene ≥97%
1-Bromo-2-methoxy-3,4-dimethylbenzene

Precaución: For Research Use Only

Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD22414742
CAS Number: 1629253-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR