7-Bromo-3-iodoimidazo[1,2-a]pyridine ≥95%

Supplier: AOBChem USA

Warning

543810-5G 543810-25G 543810-1G 543810-500MG
77635-176EA 420 USD
77635-176 77659-992 77643-638 77651-574
7-Bromo-3-iodoimidazo[1,2-a]pyridine ≥95%
7-Bromo-3-iodoimidazo[1,2-a]pyridine

Precaución: For Research Use Only

Formula: C₇H₄BrIN₂
Storage Temperature: Ambient
CAS Number: 1246184-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR