(2-Fluoro-4-(1H-pyrrol-1-yl)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

543797-1G 543797-500MG 543797-5G
77639-578EA 665 USD
77639-578 77653-012 77659-990
(2-Fluoro-4-(1H-pyrrol-1-yl)phenyl)boronic acid ≥97%
(2-Fluoro-4-(1H-pyrrol-1-yl)phenyl)boronic acid

Precaución: For Research Use Only

Formula: C₁₀H₉BFNO₂
Storage Temperature: Ambient
CAS Number: 2225179-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR