1-Bromo-2-(ethoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

543769-5G 543769-25G
77635-174EA 117 USD
77635-174 77656-470
1-Bromo-2-(ethoxymethyl)benzene ≥97%
1-Bromo-2-(ethoxymethyl)benzene

Precaución: For Research Use Only

Formula: 215.0870 g/mol
MW: C₉H₁₁BrO
Storage Temperature: Ambient
CAS Number: 80171-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR