3-Bromo-2-(methylthio)benzo[b]thiophene ≥97%

Supplier: AOBChem USA

Warning

54348-5G 54348-1G 54348-25G 54348-500MG
77635-172EA 299 USD
77635-172 77639-576 77659-988 77651-572
3-Bromo-2-(methylthio)benzo[b]thiophene ≥97%
3-Bromo-2-(methylthio)benzo[b]thiophene

Precaución: For Research Use Only

Formula: C₉H₇BrS₂
Storage Temperature: Ambient
MDL Number: MFCD28664950
CAS Number: 30489-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR