2-Bromo-N-methylnicotinamide ≥95%

Supplier: AOBChem USA

Warning

54264-5G 54264-25G 54264-1G 54264-500MG
77635-170EA 279 USD
77635-170 77656-468 77643-636 77647-578
2-Bromo-N-methylnicotinamide ≥95%
2-Bromo-N-methylnicotinamide

Precaución: For Research Use Only

Formula: 215.047 g/mol
MW: C₇H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD28154534
CAS Number: 1584140-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR