2-Iodo-4-methoxy-1-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

54224-5G 54224-1G 54224-25G 54224-500MG
77630-252EA 1484.23 USD
77630-252 77639-574 77659-986 77651-570
2-Iodo-4-methoxy-1-(methoxymethoxy)benzene ≥97%
2-Iodo-4-methoxy-1-(methoxymethoxy)benzene

Precaución: For Research Use Only

Formula: 294.086 g/mol
MW: C₉H₁₁IO₃
Storage Temperature: Ambient
MDL Number: MFCD34757266
CAS Number: 2055236-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR