2-Fluoro-3-methoxy-4-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

53850-5G 53850-1G 53850-25G 53850-500MG
77633-536EA 1381 USD
77633-536 77639-572 77659-984 77651-566
2-Fluoro-3-methoxy-4-methylpyridine ≥95%
2-Fluoro-3-methoxy-4-methylpyridine

Precaución: For Research Use Only

Formula: C₇H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD16611206
CAS Number: 1227580-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR