2-bromo-4-chloro-5-iodophenol ≥95%

Supplier: AOBChem USA

Warning

53628-5G 53628-25G 53628-1G 53628-500MG
77630-250EA 1484.23 USD
77630-250 77656-452 77643-628 77651-564
2-bromo-4-chloro-5-iodophenol ≥95%
2-bromo-4-chloro-5-iodophenol

Precaución: For Research Use Only

Formula: C₆H₃BrClIO
Storage Temperature: Ambient
MDL Number: MFCD30534476
CAS Number: 2091716-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR