Methyl 5-bromo-2-chloro-6-methylnicotinate ≥95%

Supplier: AOBChem USA

Warning

53535-5G 53535-1G 53535-25G 53535-500MG
77633-528EA 956 USD
77633-528 77639-566 77656-450 77651-560
Methyl 5-bromo-2-chloro-6-methylnicotinate ≥95%
Methyl 5-bromo-2-chloro-6-methylnicotinate

Precaución: For Research Use Only

Formula: C₈H₇BrClNO₂
MW: 264.5037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28968024
CAS Number: 1935335-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR