5-Bromo-2-fluoro-6-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

53525-5G 53525-25G 53525-1G 53525-500MG
77633-526EA 796 USD
77633-526 77656-448 77643-626 77647-566
5-Bromo-2-fluoro-6-methylnicotinaldehyde ≥97%
5-Bromo-2-fluoro-6-methylnicotinaldehyde

Precaución: For Research Use Only

Formula: 218.023 g/mol
MW: C₇H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD34757057
CAS Number: 2764730-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR