2-Chloro-3-isopropoxypyridine ≥95%

Supplier: AOBChem USA

Warning

53433-5G 53433-1G 53433-25G 53433-500MG
77633-524EA 706 USD
77633-524 77639-564 77659-976 77651-558
2-Chloro-3-isopropoxypyridine ≥95%
2-Chloro-3-isopropoxypyridine

Precaución: For Research Use Only

Formula: C₈H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD16653100
CAS Number: 1105190-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR