5-Bromo-1-fluoro-2,3-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

53367-5G 53367-1G 53367-500MG
77630-248EA 1484.23 USD
77630-248 77639-562 77651-556
5-Bromo-1-fluoro-2,3-dimethoxybenzene ≥97%
5-Bromo-1-fluoro-2,3-dimethoxybenzene

Precaución: For Research Use Only

Formula: C₈H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28641991
CAS Number: 1781895-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR