Methyl 3-bromo-2-(trifluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

53226-5G 53226-25G 53226-1G 53226-500MG
77633-522EA 755 USD
77633-522 77656-446 77643-624 77651-554
Methyl 3-bromo-2-(trifluoromethoxy)benzoate ≥97%
Methyl 3-bromo-2-(trifluoromethoxy)benzoate

Precaución: For Research Use Only

Formula: 299.041 g/mol
MW: C₉H₆BrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD28746662
CAS Number: 1805029-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR