5-Ethyl-2-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

53214-5G 53214-500MG 53214-25G 53214-1G
77633-520EA 1118 USD
77633-520 77653-010 77659-972 77643-622
5-Ethyl-2-fluorobenzaldehyde ≥95%
5-Ethyl-2-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₉FO
Storage Temperature: Ambient
MDL Number: MFCD19440182
CAS Number: 891843-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR