1-(6-Bromo-2,3-difluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

52950-5G 52950-500MG 52950-25G 52950-1G
77633-518EA 1427 USD
77633-518 77653-008 77659-970 77643-620
1-(6-Bromo-2,3-difluorophenyl)ethanone ≥95%
1-(6-Bromo-2,3-difluorophenyl)ethanone

Precaución: For Research Use Only

Formula: 235.026 g/mol
MW: C₈H₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD26679370
CAS Number: 1542563-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR