6-Bromo-N-methylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

52767-5G 52767-25G 52767-1G 52767-500MG
77635-164EA 418 USD
77635-164 77656-442 77643-618 77647-558
6-Bromo-N-methylpicolinamide ≥97%
6-Bromo-N-methylpicolinamide

Precaución: For Research Use Only

Formula: C₇H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD10699703
CAS Number: 337535-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR