1-(2,6-Difluoro-4-iodophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

52674-5G 52674-1G 52674-500MG
77630-244EA 1730.4 USD
77630-244 77643-616 77647-556
1-(2,6-Difluoro-4-iodophenyl)ethanone ≥95%
1-(2,6-Difluoro-4-iodophenyl)ethanone

Precaución: For Research Use Only

Formula: 282.026 g/mol
MW: C₈H₅F₂IO
Storage Temperature: Ambient
MDL Number: MFCD30829247
CAS Number: 1165873-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR