3-Isopropyl-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

52403-5G 52403-1G 52403-500MG
77659-966EA 2470 USD
77659-966 77643-614 77651-548
3-Isopropyl-5-methylbenzoic acid ≥95%
3-Isopropyl-5-methylbenzoic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD12547788
CAS Number: 1208077-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR