4-Bromo-1-fluoro-2-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

52342-5G 52342-1G 52342-25G 52342-500MG
77635-158EA 488 USD
77635-158 77639-552 77656-436 77647-546
4-Bromo-1-fluoro-2-naphthoic acid ≥97%
4-Bromo-1-fluoro-2-naphthoic acid

Precaución: For Research Use Only

Formula: C₁₁H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD31635910
CAS Number: 1802154-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR