(2-Isopropylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

523166-1G 523166-5G 523166-500MG
77639-548EA 630 USD
77639-548 77656-434 77651-540
(2-Isopropylphenyl)(methyl)sulfane ≥95%
(2-Isopropylphenyl)(methyl)sulphane

Precaución: For Research Use Only

Formula: 166.283 g/mol
MW: C₁₀H₁₄S
Storage Temperature: Ambient
CAS Number: 2976-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR