(2-(4-Methoxybenzyl)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

523130-5G 523130-25G 523130-1G 523130-500MG
77633-508EA 1441 USD
77633-508 77656-432 77643-606 77647-542
(2-(4-Methoxybenzyl)phenyl)boronic acid ≥95%
(2-(4-Methoxybenzyl)phenyl)boronic acid

Precaución: For Research Use Only

Formula: 242.0781 g/mol
MW: C₁₄H₁₅BO₃
Storage Temperature: Ambient
CAS Number: 2600427-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR