1-(3-Methyl-5-(methylthio)phenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

522704-5G 522704-25G 522704-1G 522704-500MG
77630-238EA 1484.23 USD
77630-238 77656-430 77643-602 77651-536
1-(3-Methyl-5-(methylthio)phenyl)cyclohexanol ≥95%
1-(3-Methyl-5-(methylthio)phenyl)cyclohexanol

Precaución: For Research Use Only

Formula: 236.373 g/mol
MW: C₁₄H₂₀OS
Storage Temperature: Ambient
CAS Number: 3037703-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR