Methyl 3-chloro-2-(1,3-dioxolan-2-yl)benzoate ≥97%

Supplier: AOBChem USA

Warning

522684-5G 522684-25G 522684-500MG 522684-1G
77630-236EA 1484.23 USD
77630-236 77636-086 77653-006 77643-600
Methyl 3-chloro-2-(1,3-dioxolan-2-yl)benzoate ≥97%
Methyl 3-chloro-2-(1,3-dioxolan-2-yl)benzoate

Precaución: For Research Use Only

Formula: 242.6556 g/mol
MW: C₁₁H₁₁ClO₄
Storage Temperature: Ambient
CAS Number: 3031785-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR