1-(4-(1,3-Dioxolan-2-yl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

522396-5G 522396-1G 522396-25G 522396-500MG
77635-154EA 558 USD
77635-154 77639-546 77656-426 77651-530
1-(4-(1,3-Dioxolan-2-yl)phenyl)ethanone ≥95%
1-(4-(1,3-Dioxolan-2-yl)phenyl)ethanone

Precaución: For Research Use Only

Formula: 192.2111 g/mol
MW: C₁₁H₁₂O₃
Storage Temperature: Ambient
CAS Number: 80463-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR