5-Bromo-2-(methylsulfonyl)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-bromo-2-methylsulphonylpyrimidine, Pyrimidine, 5-bromo-2-(methylsulphonyl)-

522395-5G 522395-25G
77635-152EA 59 USD
77635-152 77659-952
5-Bromo-2-(methylsulfonyl)pyrimidine ≥97%
5-Bromo-2-(methylsulfonyl)pyrimidine

Precaución: For Research Use Only

Formula: C₅H₅BrN₂O₂S
MW: 237.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00508247
CAS Number: 38275-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR