2-(Dimethylamino)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

522178-5G 522178-25G
77635-150EA 71 USD
77635-150 77656-424
2-(Dimethylamino)benzonitrile ≥97%
2-(Dimethylamino)benzonitrile

Precaución: For Research Use Only

Formula: C₉H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD01313389
CAS Number: 20925-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR