1-[3-Bromo-5-(trifluoromethyl)phenyl]-4-methylpiperidine ≥97%

Supplier: AOBChem USA

Warning

521399-1G 521399-5G 521399-500MG
77639-544EA 519 USD
77639-544 77656-420 77647-536
1-[3-Bromo-5-(trifluoromethyl)phenyl]-4-methylpiperidine ≥97%
1-[3-Bromo-5-(trifluoromethyl)phenyl]-4-methylpiperidine

Precaución: For Research Use Only

Formula: C₁₃H₁₅BrF₃N
Storage Temperature: Ambient
CAS Number: 1704068-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR