2,5-Dimethylbenzophenone ≥95%

Supplier: AOBChem USA

Warning

520640-5G 520640-1G 520640-25G 520640-500MG
77635-148EA 473 USD
77635-148 77639-542 77656-418 77651-528
2,5-Dimethylbenzophenone ≥95%
2,5-Dimethylbenzophenone

Precaución: For Research Use Only

Formula: C₁₅H₁₄O
Storage Temperature: Ambient
CAS Number: 4044-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR