(3-Methoxy-5-(1-methyl-1H-pyrazol-4-yl)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

520637-1G 520637-5G 520637-500MG
77635-146EA 1158 USD
77635-146 77656-416 77647-534
(3-Methoxy-5-(1-methyl-1H-pyrazol-4-yl)phenyl)boronic acid ≥97%
(3-Methoxy-5-(1-methyl-1H-pyrazol-4-yl)phenyl)boronic acid

Precaución: For Research Use Only

Formula: 232.0435 g/mol
MW: C₁₁H₁₃BN₂O₃
Storage Temperature: Ambient
CAS Number: 1373043-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR