Methyl 3-fluoro-2,6-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

52041-1G 52041-5G 52041-500MG
77635-144EA 817 USD
77635-144 77656-414 77647-530
Methyl 3-fluoro-2,6-dimethylbenzoate ≥97%
Methyl 3-fluoro-2,6-dimethylbenzoate

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD28794300
CAS Number: 26829-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR