(3-Fluoro-2,6-dimethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

52038-5G 52038-1G 52038-500MG
77656-412EA 2282 USD
77656-412 77643-592 77647-528
(3-Fluoro-2,6-dimethylphenyl)boronic acid ≥97%
(3-Fluoro-2,6-dimethylphenyl)boronic acid

Precaución: For Research Use Only

Formula: 167.973 g/mol
MW: C₈H₁₀BFO₂
Storage Temperature: Ambient
MDL Number: MFCD23712907
CAS Number: 2950233-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR