2,3,4-Tribromophenol ≥97%

Supplier: AOBChem USA

Warning

519339-1G 519339-500MG 519339-5G
77635-142EA 935 USD
77635-142 77652-996 77656-402
2,3,4-Tribromophenol ≥97%
2,3,4-Tribromophenol

Precaución: For Research Use Only

Formula: C₆H₃Br₃O
Storage Temperature: Ambient
MDL Number: MFCD09037673
CAS Number: 138507-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR