Methyl 2-(2-chloro-5-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

519200-5G 519200-25G 519200-1G 519200-500MG
77630-208EA 1512.04 USD
77630-208 77636-080 77643-574 77651-510
Methyl 2-(2-chloro-5-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Methyl 2-(2-chloro-5-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 266.601 g/mol
MW: C₁₀H₆ClF₃O₃
Storage Temperature: Ambient
CAS Number: 2095410-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR