Phenyl(2,4,6-trifluorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

519198-5G 519198-1G 519198-500MG
77659-930EA 2001 USD
77659-930 77643-572 77651-508
Phenyl(2,4,6-trifluorophenyl)methanone ≥95%
Phenyl(2,4,6-trifluorophenyl)methanone

Precaución: For Research Use Only

Formula: 236.189 g/mol
MW: C₁₃H₇F₃O
Storage Temperature: Ambient
CAS Number: 243448-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR