Ethyl 2-(2-chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

519065-5G 519065-25G 519065-1G 519065-500MG
77630-204EA 1484.23 USD
77630-204 77636-078 77639-524 77651-504
Ethyl 2-(2-chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₁H₇ClF₄O₃
Storage Temperature: Ambient
CAS Number: 3037703-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR