(3-Bromo-2-fluoro-6-iodophenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

519058-5G 519058-25G 519058-1G 519058-500MG
77633-502EA 1441 USD
77633-502 77656-392 77643-566 77647-514
(3-Bromo-2-fluoro-6-iodophenyl)(cyclopropyl)methanol ≥95%
(3-Bromo-2-fluoro-6-iodophenyl)(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: 370.985 g/mol
MW: C₁₀H₉BrFIO
Storage Temperature: Ambient
CAS Number: 3037705-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR