1-(7-Methylbenzo[b]thiophen-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

519033-1G 519033-5G 519033-500MG
77635-140EA 1132 USD
77635-140 77659-928 77651-498
1-(7-Methylbenzo[b]thiophen-2-yl)ethanone ≥95%
1-(7-Methylbenzo[b]thiophen-2-yl)ethanone

Precaución: For Research Use Only

Formula: 190.262 g/mol
MW: C₁₁H₁₀OS
Storage Temperature: Ambient
CAS Number: 1505-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR