7-Methyl-2-(methylthio)benzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

519029-5G 519029-25G 519029-1G 519029-500MG
77630-194EA 1484.23 USD
77630-194 77636-074 77647-512 77651-496
7-Methyl-2-(methylthio)benzo[b]thiophene ≥95%
7-Methyl-2-(methylthio)benzo[b]thiophene

Precaución: For Research Use Only

Formula: 194.316 g/mol
MW: C₁₀H₁₀S₂
Storage Temperature: Ambient
CAS Number: 1824310-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR