Ethyl 2-(3-bromo-2-fluoro-6-methoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

519012-1G 519012-5G 519012-500MG
77639-516EA 619 USD
77639-516 77659-924 77651-492
Ethyl 2-(3-bromo-2-fluoro-6-methoxyphenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-bromo-2-fluoro-6-methoxyphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 305.097 g/mol
MW: C₁₁H₁₀BrFO₄
Storage Temperature: Ambient
CAS Number: 1892044-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR