Tert-butyl 3-bromo-2-fluoro-6-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

519011-5G 519011-25G 519011-1G 519011-500MG
77630-192EA 1823.1 USD
77630-192 77636-072 77639-514 77652-992
Tert-butyl 3-bromo-2-fluoro-6-methoxybenzoate ≥95%
Formula: C₁₂H₁₄BrFO₃
Storage Temperature: Ambient
CAS Number: 3037696-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR