2-(3-Bromo-2-fluoro-6-methoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

519010-5G 519010-1G 519010-500MG
77659-922EA 1992 USD
77659-922 77643-562 77647-510
2-(3-Bromo-2-fluoro-6-methoxyphenyl)propan-2-ol ≥95%
2-(3-Bromo-2-fluoro-6-methoxyphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₀H₁₂BrFO₂
Storage Temperature: Ambient
CAS Number: 1780887-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR