Tert-butyl 2-fluoro-3,6-dimethoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

519005-5G 519005-25G 519005-1G 519005-500MG
77630-186EA 1484.23 USD
77630-186 77636-070 77643-558 77647-506
Tert-butyl 2-fluoro-3,6-dimethoxybenzoate ≥95%
Tert-butyl 2-fluoro-3,6-dimethoxybenzoate

Precaución: For Research Use Only

Formula: C₁₃H₁₇FO₄
Storage Temperature: Ambient
CAS Number: 3037705-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR