2-(4-Fluorophenoxy)-N-methoxy-N-methylacetamide ≥97%

Supplier: AOBChem USA

Warning

518995-5G 518995-1G 518995-25G 518995-500MG
77633-498EA 1381 USD
77633-498 77639-506 77659-910 77647-496
2-(4-Fluorophenoxy)-N-methoxy-N-methylacetamide ≥97%
2-(4-Fluorophenoxy)-N-methoxy-N-methylacetamide

Precaución: For Research Use Only

Formula: 213.2055 g/mol
MW: C₁₀H₁₂FNO₃
Storage Temperature: Ambient
CAS Number: 524713-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR