5-Bromo-2-chloro-3-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

518921-5G 518921-25G 518921-1G 518921-500MG
77630-172EA 1484.23 USD
77630-172 77636-066 77643-548 77647-494
5-Bromo-2-chloro-3-fluorobenzamide ≥97%
5-Bromo-2-chloro-3-fluorobenzamide

Precaución: For Research Use Only

Formula: 252.468 g/mol
MW: C₇H₄BrClFNO
Storage Temperature: Ambient
CAS Number: 2091468-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR