(3,5-Dichloro-4-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51776-5G 51776-1G 51776-25G
77635-138EA 228 USD
77635-138 77639-500 77656-378
(3,5-Dichloro-4-iodophenyl)(methyl)sulfane ≥97%
(3,5-Dichloro-4-iodophenyl)(methyl)sulphane

Precaución: For Research Use Only

Formula: C₇H₅Cl₂IS
Storage Temperature: Ambient
MDL Number: MFCD34530145
CAS Number: 2706294-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR