5-Chloro-6-methoxy-2-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

51655-5G 51655-1G 51655-500MG 51655-25G
77633-494EA 886 USD
77633-494 77639-498 77652-988 77659-908
5-Chloro-6-methoxy-2-naphthoic acid ≥97%
5-Chloro-6-methoxy-2-naphthoic acid

Precaución: For Research Use Only

Formula: 236.651 g/mol
MW: C₁₂H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD19303025
CAS Number: 408327-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR