2,2,2-Trifluoro-N-(3-fluoro-2-iodo-4-methoxyphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

51636-5G 51636-25G 51636-1G 51636-500MG
77635-136EA 488 USD
77635-136 77656-376 77643-546 77647-488
2,2,2-Trifluoro-N-(3-fluoro-2-iodo-4-methoxyphenyl)acetamide ≥97%
2,2,2-Trifluoro-N-(3-fluoro-2-iodo-4-methoxyphenyl)acetamide

Precaución: For Research Use Only

Formula: 363.048 g/mol
MW: C₉H₆F₄INO₂
Storage Temperature: Ambient
MDL Number: MFCD32902963
CAS Number: 2586127-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR