1-(3-(Cyclopropylmethoxy)-4-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

515051-5G 515051-1G 515051-500MG
77630-166EA 1484.23 USD
77630-166 77639-496 77647-482
1-(3-(Cyclopropylmethoxy)-4-methylphenyl)cyclobutanol ≥95%
1-(3-(Cyclopropylmethoxy)-4-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₅H₂₀O₂
Storage Temperature: Ambient
CAS Number: 3037703-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR